missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF2AK2 Protein
A DNA sequence encoding the Homo sapiens (Human) EIF2AK2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5969-ye-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001129123.1 | |
EIF2AK2 Protein | |
Research Use Only | |
EIF2AK2 | |
Recombinant Protein | |
Yeast | |
AGDLSAGFFM EELNTYRQKQ GVVLKYQELP NSGPPHDRRF TFQVIIDGRE FPEGEGRSKK EAKNAAAKLA VEILNKEKKA VSPLLLTTTN SSEGLSMGNY IGLINRIAQK KRLTVNYEQC ASGVHGPEGF HYKCKMGQKE YSIGTGSTKQ EAKQLAAKLA YLQILSEETS VKSDYLSSGS FATTCESQSN SLVTSTLASE SSSEGDFSAD TSEINSNSDS LNSSSLLMNG LRNNQRKAKR SLAPRFDLPD MKETKYTVDK RFGMDFKEIE LIGSGGFGQV FKAKHRIDGK TYVIKRVKYN NEKAEREVKA LAKLDHVNIV HYNGCWDGFD YDPETSDDSL ESSDYDPENS KNSSRSKTKC LFIQMEFCDK GTLEQWIEKR RGEKLDKVLA LELFEQITKG VDYIHSKKLI HRDLKPSNIF LVDTKQVKIG DFGLVTSLKN DGKRTRSKGT LRYMSPEQIS SQDYGKEVDL YALGLILAEL LHVCDTAFET SKFFTDLRDG IISDIFDKKE KTLLQKLLSK KPEDRPNTSE ILRTLTVWKK SPEKNERHTC | |
Greater than 90% as determined by SDS-PAGE. |
5610 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |