missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EGFL6 Protein
A cDNA sequence encoding the EGFL6 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11758-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
EGFL6 Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The EGFL6 protein was lyophilized from a 0.2microm filtered solution in 20mM MES and 500mM NaCl, pH 6.0. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
HEK 293 | |
MPLPWSLALP LLLSWVAGGF GNAASARHHG LLASARQPGV CHYGTKLACC YGWRRNSKGV CEATCEPGCK FGECVGPNKC RCFPGYTGKT CSQDVNECGM KPRPCQHRCV NTHGSYKCFC LSGHMLMPDA TCVNSRTCAM INCQYSCEDT EEGPQCLCPS SGLRLAPNGR DCLDIDECAS GKVICPYNRR CVNTFGSYYC KCHIGFELQY ISGRYDCIDI NECTMDSHTC SHHANCFNTQ GSFKCKCKQG YKGNGLRCSA IPENSVKEVL RAPGTIKDRI KKLLAHKNSM KKKAKIKNVT PEPTRTPTPK VNLQPFNYEE IVSRGGNSHG GKKGNEEKMK EGLEDEKREE KALKNDIEER SLRGDVFFPK VNEAGEFGLI LVQRKALTSK LEHKDLNISV DCSFNHGICD WKQDREDDFD WNPADRDNAI GFYMAVPALA GHKKDIGRLK LLLPDLQPQS NFCLLFDYRL AGDKVGKLRV FVKNSNNALA WEKTTSEDEK WKTGKIQLYQ GTDATKSIIF EAERGKGKTG EIAVDGVLLV SGLCPDSLLS VDD |