missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DUSP13, isoform A Protein
A DNA sequence encoding the Homo sapiens (Human) DUSP13, isoform A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7728-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001007272.1 | |
DUSP13, isoform A Protein | |
Research Use Only | |
DUSP13 | |
Recombinant Protein | |
E. coli | |
MAETSLPELG GEDKATPCPS ILELEELLRA GKSSCSRVDE VWPNLFIGDA ATANNRFELW KLGITHVLNA AHKGLYCQGG PDFYGSSVSY LGVPAHDLPD FDISAYFSSA ADFIHRALNT PGAKVLVHCV VGVSRSATLV LAYLMLHQRL SLRQAVITVR QHRWVFPNRG FLHQLCRLDQ QLRGAGQS | |
Greater than 90% as determined by SDS-PAGE. |
51207 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |