missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DTW domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) DTW domain-containing protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5953-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001138427.1 | |
DTW domain-containing protein 1 | |
Research Use Only | |
DTWD1 | |
Recombinant Protein | |
E. coli | |
MSLNPPIFLK RSEENSSKFV ETKQSQTTSI ASEDPLQNLC LASQEVLQKA QQSGRSKCLK CGGSRMFYCY TCYVPVENVP IEQIPLVKLP LKIDIIKHPN ETDGKSTAI | |
Greater than 90% as determined by SDS-PAGE. |
56986 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |