missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DR6 / TNFRSF21 Protein
A DNA sequence encoding the Homo sapiens (Human) DR6 / TNFRSF21, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6815-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_055267.1 | |
DR6 / TNFRSF21 Protein | |
Research Use Only | |
TNFRSF21 | |
Recombinant Protein | |
E. coli | |
RKSSRTLKKG PRQDPSAIVE KAGLKKSMTP TQNREKWIYY CNGHGIDILK LVAAQVGSQW KDIYQFLCNA SEREVAAFSN GYTADHERAY AALQHWTIRG PEASLAQLIS ALRQHRRNDV VEKIRGLMED TTQLETDKLA LPMSPSPLSP SPIPSPNAKL ENSALLTVEP SPQDKNKGFF VDESEPLLRC DSTSSGSSAL SRNGSFITKE KKDTVLRQVR LDPCDLQPIF DDMLHFLNPE ELRVIEEIPQ AEDKLDRLFE IIGVKSQEAS QTLLDSVYSH LPDLL | |
Greater than 90% as determined by SDS-PAGE. |
27242 | |
10 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |