missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNTTIP1 Protein
A DNA sequence encoding the Homo sapiens (Human) DNTTIP1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5942-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_443183.1 | |
DNTTIP1 Protein | |
Research Use Only | |
DNTTIP1 | |
Recombinant Protein | |
E. coli | |
MGATGDAEQP RGPSGAERGG LELGDAGAAG QLVLTNPWNI MIKHRQVQRR GRRSQMTTSF TDPAISMDLL RAVLQPSINE EIQTVFNKYM KFFQKAALNV RDNVGEEVDA EQLIQEACRS CLEQAKLLFS DGEKVIPRLT HELPGIKRGR QAEEECAHRG SPLPKKRKGR PPGHILSSDR AAAGMVWKPK SCEPIRREGP KWDPARLNES TTFVLGSRAN KALGMGGTRG RIYIKHPHLF KYAADPQDKH WLAEQHHMRA TGGKMAYLLI EEDIRDLAAS DDYRGCLDLK LEELKSFVLP SWMVEKMRKY METLRTENEH RAVEAPPQT | |
Greater than 90% as determined by SDS-PAGE. |
116092 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |