missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA-binding protein inhibitor ID-1 Protein
A DNA sequence encoding the Homo sapiens (Human) DNA-binding protein inhibitor ID-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6190-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002156.2 | |
DNA-binding protein inhibitor ID-1 Protein | |
Research Use Only | |
ID1 | |
Recombinant Protein | |
E. coli | |
MKVASGSTAT AAAGPSCALK AGKTASGAGE VVRCLSEQSV AISRCAGGAG ARLPALLDEQ QVNVLLYDMN GCYSRLKELV PTLPQNRKVS KVEILQHVID YIRDLQLELN SESEVGTPGG RGLPVRAPLS TLNGEISALT AEAACVPADD RILCR | |
Greater than 90% as determined by SDS-PAGE. |
3397 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |