missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dihydropyrimidinase-related protein 1
A DNA sequence encoding the Homo sapiens (Human) Dihydropyrimidinase-related protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7611-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001014809.1 | |
Dihydropyrimidinase-related protein 1 | |
Research Use Only | |
CRMP1 | |
Recombinant Protein | |
E. coli | |
MSYQGKKSIP HITSDRLLIK GGRIINDDQS LYADVYLEDG LIKQIGENLI VPGGVKTIEA NGRMVIPGGI DVNTYLQKPS QGMTAADDFF QGTRAALVGG TTMIIDHVVP EPGSSLLTSF EKWHEAADTK SCCDYSLHVD ITSWYDGVRE ELEVLVQDKG VNSFQVYMAY KDVYQMSDSQ LYEAFTFLKG LGAVILVHAE NGDLIAQEQK RILEMGITGP EGHALSRPEE LEAEAVFRAI TIAGRINCPV YITKVMSKSA ADIIALARKK GPLVFGEPIA ASLGTDGTHY WSKNWAKAAA FVTSPPLSPD PTTPDYLTSL LACGDLQVTG SGHCPYSTAQ KAVGKDNFTL IPEGVNGIEE RMTVVWDKAV ATGKMDENQF VAVTSTNAAK IFNLYPRKGR IAVGSDADVV IWDPDKLKTI TAKSHKSAVE YNIFEGMECH GSPLVVISQG KIVFEDGNIN VNKGMGRFIP RKAFPEHLYQ RVKIRNKVFG LQGVSRGMYD GPVYEVPATP KYATPAPSAK SSPSKHQPPP IRNLHQSNFS LSGAQIDDNN PRRTGHRIVA PPGGRSNITS LG | |
Greater than 90% as determined by SDS-PAGE. |
1400 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |