missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dickkopf-related protein 1
A DNA sequence encoding the Homo sapiens (Human) Dickkopf-related protein 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8081-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_055236.1 | |
Dickkopf-related protein 1 | |
Research Use Only | |
DKK2 | |
Recombinant Protein | |
E. coli | |
KLNSIKSSLG GETPGQAANR SAGMYQGLAF GGSKKGKNLG QAYPCSSDKE CEVGRYCHSP HQGSSACMVC RRKKKRCHRD GMCCPSTRCN NGICIPVTES ILTPHIPALD GTRHRDRNHG HYSNHDLGWQ NLGRPHTKMS HIKGHEGDPC LRSSDCIEGF CCARHFWTKI CKPVLHQGEV CTKQRKKGSH GLEIFQRCDC AKGLSCKVWK DATYSSKARL HVCQKI | |
Greater than 90% as determined by SDS-PAGE. |
27123 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |