missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DECR2 Protein
A cDNA sequence encoding the DECR2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11616-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
DECR2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAQPPPD VEGDDCLPAY RHLFCPDLLR DKVAFITGGG SGIGFRIAEI FMRHGCHTVI ASRSLPRVLT AARKLAGATG RRCLPLSMDV RAPPAVMAAV DQALKEFGRI DILINCAAGN FLCPAGALSF NAFKTVMDID TSGTFNVSRV LYEKFFRDHG GVIVNITATL GNRGQALQVH AGSAKAAVDA MTRHLAVEWG PQNIRVNSLA PGPISGTEGL RRLGGPQASL STKVTASPLQ RLGNKTEIAH SVLYLASPLA SYVTGAVLVA DGGAWLTFPN GVKGLPDFAS FSAKL | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DECR2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 40% glycerol, 0.15M NaCl and 1mM DTT. |