missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DCX Protein
A DNA sequence encoding the Homo sapiens (Human) DCX, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5920-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000546.2 | |
DCX Protein | |
Research Use Only | |
DCX | |
Recombinant Protein | |
E. coli | |
MKTLPLHSHC TEMQRLLPKL EMLTLGSSFC SLQGEFCQAM DSFTTVSHVG MCEETDASFN VFSPKFQFDR SHCQSLRFHQ NMELDFGHFD ERDKTSRNMR GSRMNGLPSP THSAHCSFYR TRTLQALSNE KKAKKVRFYR NGDRYFKGIV YAVSSDRFRS FDALLADLTR SLSDNINLPQ GVRYIYTIDG SRKIGSMDEL EEGESYVCSS DNFFKKVEYT KNVNPNWSVN VKTSANMKAP QSLASSNSAQ ARENKDFVRP KLVTIIRSGV KPRKAVRVLL NKKTAHSFEQ VLTDITEAIK LETGVVKKLY TLDGKQVTCL HDFFGDDDVF IACGPEKFRY AQDDFSLDEN ECRVMKGNPS ATAGPKASPT PQKTSAKSPG PMRRSKSPAD SANGTSSSQL STPKSKQSPI STPTSPGSLR KHKDLYLPLS LDDSDSLGDS M | |
Greater than 90% as determined by SDS-PAGE. |
1641 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |