missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DCK Protein
A cDNA sequence encoding the DCK was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11599-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
DCK Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMATP PKRSCPSFSA SSEGTRIKKI SIEGNIAAGK STFVNILKQL CEDWEVVPEP VARWCNVQST QDEFEELTMS QKNGGNVLQM MYEKPERWSF TFQTYACLSR IRAQLASLNG KLKDAEKPVL FFERSVYSDR YIFASNLYES ECMNETEWTI YQDWHDWMNN QFGQSLELDG IIYLQATPET CLHRIYLRGR NEEQGIPLEY LEKLHYKHES WLLHRTLKTN FDYLQEVPIL TLDVNEDFKD KYESLVEKVK EFLSTL | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DCK solution (0.5 mg/ml) contains 20mM Tris-HCl Buffer (pH 7.5), 1mM DTT, 0.1mM PMSF, 2mM EDTA and 10% Glycerol. |