missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytokeratin, Acidic Protein
A DNA sequence encoding the Homo sapiens (Human) Cytokeratin, Acidic, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7778-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_778253.2 | |
Cytokeratin, Acidic Protein | |
Research Use Only | |
KRT77 | |
Recombinant Protein | |
E. coli | |
MSHQFSSQSA FSSMSRRVYS TSSSAGSGGG SPAVGSVCYA RGRCGGGGYG IHGRGFGSRS LYNLGGSRSI SINLMGRSTS GFCQGGGVGG FGGGRGFGVG STGAGGFGGG GFGGAGFGTS NFGLGGFGPY CPPGGIQEVT INQSLLEPLH LEVDPEIQRI KTQEREQIMV LNNKFASFID KVRFLEQQNQ VLQTKWELLQ QVNTSTGTNN LEPLLENYIG DLRRQVDLLS AEQMRQNAEV RSMQDVVEDY KSKYEDEINK RTGSENDFVV LKKDVDAAYV SKVDLESRVD TLTGEVNFLK YLFLTELSQV QTHISDTNVI LSMDNNRSLD LDSIIDAVRT QYELIAQRSK DEAEALYQTK YQELQITAGR HGDDLKNSKM EIAELNRTVQ RLQAEISNVK KQIEQMQSLI SDAEERGEQA LQDAWQKLQD LEEALQQSKE ELARLLRDYQ AMLGVKLSLD VEIATYRQLL EGEESRMSGE LQSHVSISVQ NSQVSVNGGA GGGGSYGSGG YGGGSGGGYG GGRSYRGGGA RGRSGGGYGS GCGGGGGSYG GSGRSGRGSS RVQIIQTSTN TSHRRILE | |
Greater than 90% as determined by SDS-PAGE. |
374454 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |