missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytokeratin 18 Protein
A DNA sequence encoding the Homo sapiens (Human) Cytokeratin 18, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6278-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000215.1 | |
Cytokeratin 18 Protein | |
Research Use Only | |
KRT18 | |
Recombinant Protein | |
E. coli | |
TNYRSLGSVQ APSYGARPVS SAASVYAGAG GSGSRISVSR STSFRGGMGS GGLATGIAGG LAGMGGIQNE KETMQSLNDR LASYLDRVRS LETENRRLES KIREHLEKKG PQVRDWSHYF KIIEDLRAQI FANTVDNARI VLQIDNARLA ADDFRVKYET ELAMRQSVEN DIHGLRKVID DTNITRLQLE TEIEALKEEL LFMKKNHEEE VKGLQAQIAS SGLTVEVDAP KSQDLAKIMA DIRAQYDELA RKNREELDKY WSQQIEESTT VVTTQSAEVG AAETTLTELR RTVQSLEIDL DSMRNLKASL ENSLREVEAR YALQMEQLNG ILLHLESELA QTRAEGQRQA QEYEALLNIK VKLEAEIATY RRLLEDGEDF NLGDALDSSN SMQTIQKTTT RRIVDGKVVS ETNDTKVLRH | |
Greater than 90% as determined by SDS-PAGE. |
3875 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |