missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytoglobin / CYGB Protein
A DNA sequence encoding the Homo sapiens (Human) Cytoglobin / CYGB, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5901-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_599030.1 | |
Cytoglobin / CYGB Protein | |
Research Use Only | |
CYGB | |
Recombinant Protein | |
E. coli | |
MEKVPGEMEI ERRERSEELS EAERKAVQAM WARLYANCED VGVAILVRFF VNFPSAKQYF SQFKHMEDPL EMERSPQLRK HACRVMGALN TVVENLHDPD KVSSVLALVG KAHALKHKVE PVYFKILSGV ILEVVAEEFA SDFPPETQRA WAKLRGLIYS HVTAAYKEVG WVQQVPNATT PPATLPSSGP | |
Greater than 90% as determined by SDS-PAGE. |
114757 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |