missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytochrome P450 11B2, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Cytochrome P450 11B2, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5903-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000489.3 | |
Cytochrome P450 11B2, mitochondrial Protein | |
Research Use Only | |
CYP11B2 | |
Recombinant Protein | |
E. coli | |
GTRAARAPRT VLPFEAMPQH PGNRWLRLLQ IWREQGYEHL HLEMHQTFQE LGPIFRYNLG GPRMVCVMLP EDVEKLQQVD SLHPCRMILE PWVAYRQHRG HKCGVFLLNG PEWRFNRLRL NPDVLSPKAV QRFLPMVDAV ARDFSQALKK KVLQNARGSL TLDVQPSIFH YTIEASNLAL FGERLGLVGH SPSSASLNFL HALEVMFKST VQLMFMPRSL SRWISPKVWK EHFEAWDCIF QYGDNCIQKI YQELAFNRPQ HYTGIVAELL LKAELSLEAI KANSMELTAG SVDTTAFPLL MTLFELARNP DVQQILRQES LAAAASISEH PQKATTELPL LRAALKETLR LYPVGLFLER VVSSDLVLQN YHIPAGTLVQ VFLYSLGRNA ALFPRPERYN PQRWLDIRGS GRNFHHVPFG FGMRQCLGRR LAEAEMLLLL HHVLKHFLVE TLTQEDIKMV YSFILRPGTS PLLTFRAIN | |
Greater than 90% as determined by SDS-PAGE. |
1585 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |