missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cyclophilin A / PPIA / CYPA Protein
A DNA sequence encoding the Homo sapiens (Human) Cyclophilin A / PPIA / CYPA, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8588-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001287910.1 | |
Cyclophilin A / PPIA / CYPA Protein | |
Research Use Only | |
PPIA | |
Recombinant Protein | |
E. coli | |
VNPTVFFDIA VDGEPLGRVS FELFADKVPK TAENFRALST GEKGFGYKGS CFHRIIPGFM CQGGDFTRHN GTGGKSIYGE KFEDENFILK HTGPGILSMA NAGPNTNGSQ FFICTAKTEW LDGKHVVFGK VKEGMNIVEA MERFGSRNGK TSKKITIADC GQLE | |
Greater than 90% as determined by SDS-PAGE. |
5478 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
Untagged | |
Tris-based buffer, 50% glycerol |