missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL5 / ENA-78 Protein
A DNA sequence encoding the Homo sapiens (Human) CXCL5 / ENA-78, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8535-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002985.1 | |
CXCL5 / ENA-78 Protein | |
Research Use Only | |
CXCL5 | |
Recombinant Protein | |
E. coli | |
AGPAAAVLRE LRCVCLQTTQ GVHPKMISNL QVFAIGPQCS KVEVVASLKN GKEICLDPEA PFLKKVIQKI LDGG | |
Greater than 90% as determined by SDS-PAGE. |
6374 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |