missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CUEDC1 Protein
A cDNA sequence encoding the CUEDC1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11566-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CUEDC1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTSLFRR SSSGSGGGGT AGARGGGGGT AAPQELNNSR PARQVRRLEF NQAMDDFKTM FPNMDYDIIE CVLRANSGAV DATIDQLLQM NLEGGGSSGG VYEDSSDSED SIPPEILERT LEPDSSDEEP PPVYSPPAYH MHVFDRPYPL APPTPPPRID ALGSGAPTSQ RRYRNWNPPL LGNLPDDFLR ILPQQLDSIQ GNAGGPKPGS GEGCPPAMAG PGPGDQESRW KQYLEDERIA LFLQNEEFMK ELQRNRDFLL ALERDRLKYE SQKSKSSSVA VGNDFGFSSP VPGTGDANPA VSEDALFRDK LKHMGKSTRR KLFELARAFS EKTKMRKSKR KHLLKHQSLG AAASTANLLD DVEGHACDED FRGRRQEAPK VEEGLREGQ | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CUEDC1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |