missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CSN2 Protein
A cDNA sequence encoding the CSN2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11531-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CSN2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMRETIE SLSSSEESIT EYKQKVEKVK HEDQQQGEDE HQDKIYPSFQ PQPLIYPFVE PIPYGFLPQN ILPLAQPAVV LPVPQPEIME VPKAKDTVYT KGRVMPVLKS PTIPFFDPQI PKLTDLENLH LPLPLLQPLM QQVPQPIPQT LALPPQPLWSVPQPKVLPIP QQVVPYPQRA VPVQALLLNQ ELLLNPTHQI YPVTQPLAPV HNPISV | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CSN2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.5M NaCl, 0.25M imidazole, 1mM DTT and 0.1mM PMSF. |