missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRABP1 / RBP5 Protein
A DNA sequence encoding the Homo sapiens (Human) CRABP1 / RBP5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5864-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004369.1 | |
CRABP1 / RBP5 Protein | |
Research Use Only | |
CRABP1 | |
Recombinant Protein | |
E. coli | |
PNFAGTWKMR SSENFDELLK ALGVNAMLRK VAVAAASKPH VEIRQDGDQF YIKTSTTVRT TEINFKVGEG FEEETVDGRK CRSLPTWENE NKIHCTQTLL EGDGPKTYWT RELANDELIL TFGADDVVCT RIYVRE | |
Greater than 90% as determined by SDS-PAGE. |
1381 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |