Learn More
enQuireBio™ Recombinant Human COX4NB Protein
A cDNA sequence encoding the COX4NB was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11491-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
COX4NB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPGVKLTTQA YCKMVLHGAK YPHCAVNGLL VAEKQKPRKE HLPLGGPGAH HTLFVDCIPL FHGTLALAPM LEVALTLIDS WCKDHSYVIA GYYQANERVK DASPNQVAEK VASRIAEGFS DTALIMVDNT KFTMDCVAPT IHVYEHHENR WRCRDPHHDY CEDWPEAQRI SASLLDSRSY ETLVDFDNHL DDIRNDWTNP EINKAVLHLC | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The COX4NB solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 10% glycerol. |