missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Complement C1q subcomponent subunit A Protein
A DNA sequence encoding the Homo sapiens (Human) Complement C1q subcomponent subunit A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5734-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001334394.1 | |
Complement C1q subcomponent subunit A Protein | |
Research Use Only | |
C1QA | |
Recombinant Protein | |
E. coli | |
EDLCRAPDGK KGEAGRPGRR GRPGLKGEQG EPGAPGIRTG IQGLKGDQGE PGPSGNPGKV GYPGPSGPLG ARGIPGIKGT KGSPGNIKDQ PRPAFSAIRR NPPMGGNVVI FDTVITNQEE PYQNHSGRFV CTVPGYYYFT FQVLSQWEIC LSIVSSSRGQ VRRSLGFCDT TNKGLFQVVS GGMVLQLQQG DQVWVEKDPK KGHIYQGSEA DSVFSGFLIF PSA | |
Greater than 90% as determined by SDS-PAGE. |
712 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |