missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COMMD6 Protein
A cDNA sequence encoding the COMMD6 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11480-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
COMMD6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEASSEP PLDAKSDVTN QLVDFQWKLG MAVSSDTCRS LKYPYVAVML KVADHSGQVK TKCFEMTIPQ FQNFYRQFKE IAAVIETV | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The COMMD6 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.15M NaCl and 1mM DTT. |