missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COMM domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) COMM domain-containing protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7824-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_689729.1 | |
COMM domain-containing protein 1 | |
Research Use Only | |
COMMD1 | |
Recombinant Protein | |
E. coli | |
AAGELEGGKP LSGLLNALAQ DTFHGYPGIT EELLRSQLYP EVPPEEFRPF LAKMRGILKS IASADMDFNQ LEAFLTAQTK KQGGITSDQA AVISKFWKSH KTKIRESLMN QSRWNSGLRG LSWRVDGKSQ SRHSAQIHTP VAIIELELGK YGQESEFLCL EFDEVKVNQI LKTLSEVEES ISTLISQPN | |
Greater than 80% as determined by SDS-PAGE. |
150684 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |