missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COL4A3 Protein
A DNA sequence encoding the Homo sapiens (Human) COL4A3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5853-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000082.2 | |
COL4A3 Protein | |
Research Use Only | |
COL4A3 | |
Recombinant Protein | |
E. coli | |
FVFTRHSQTT AIPSCPEGTV PLYSGFSFLF VQGNQRAHGQ DLGTLGSCLQ RFTTMPFLFC NVNDVCNFAS RNDYSYWLST PALMPMNMAP ITGRALEPYI SRCTVCEGPA IAIAVHSQTT DIPPCPHGWI SLWKGFSFIM FTSAGSEGTG QALASPGSCL EEFRASPFLE CHGRGTCNYY SNSYSFWLAS LNPERMFRKP IPSTVKAGEL EKIISRCQVC MK | |
Greater than 90% as determined by SDS-PAGE. |
1285 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |