missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CNTF Protein
Recombinant Protein
Marca: enQuireBio™ QP5371-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000605.1 | |
CNTF Protein | |
Research Use Only | |
CNTF | |
Recombinant Protein | |
E. coli | |
MAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM | |
Greater than 95% as determined by SDS-PAGE. |
1270 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 325 ng/mL in the TF-1 cell proliferation assay. The bioactivity acceptance criteria is set at 3.1 x 10^3 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |