missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CNDP2 / CPGL / PEPA Protein
A DNA sequence encoding the Homo sapiens (Human) CNDP2 / CPGL / PEPA, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8005-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001161971.1 | |
CNDP2 / CPGL / PEPA Protein | |
Research Use Only | |
CNDP2 | |
Recombinant Protein | |
E. coli | |
AALTTLFKYI DENQDRYIKK LAKWVAIQSV SAWPEKRGEI RRMMEVAAAD VKQLGGSVEL VDIGKQKLPD GSEIPLPPIL LGRLGSDPQK KTVCIYGHLD VQPAALEDGW DSEPFTLVER DGKLYGRGST DDKGPVAGWI NALEAYQKTG QEIPVNVRFC LEGMEESGSE GLDELIFARK DTFFKDVDYV CISDNYWLGK KKPCITYGLR GICYFFIEVE CSNKDLHSGV YGGSVHEAMT DLILLMGSLV DKRGNILIPG INEAVAAVTE EEHKLYDDID FDIEEFAKDV GAQILLHSHK KDILMHRWRY PSLSLHGIEG AFSGSGAKTV IPRKVVGKFS IRLVPNMTPE VVGEQVTSYL TKKFAELRSP NEFKVYMGHG GKPWVSDFSH PHYLAGRRAM KTVFGVEPDL TREGGSIPVT LTFQEATGKN VMLLPVGSAD DGAHSQNEKL NRYNYIEGTK MLAAYLYEVS QLKD | |
Greater than 90% as determined by SDS-PAGE. |
55748 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |