missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Chymotrypsin-like elastase family member 3A Protein
A DNA sequence encoding the Homo sapiens (Human) Chymotrypsin-like elastase family member 3A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5979-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005738.4 | |
Chymotrypsin-like elastase family member 3A Protein | |
Research Use Only | |
CELA3A | |
Recombinant Protein | |
E. coli | |
VVHGEDAVPY SWPWQVSLQY EKSGSFYHTC GGSLIAPDWV VTAGHCISRD LTYQVVLGEY NLAVKEGPEQ VIPINSEELF VHPLWNRSCV ACGNDIALIK LSRSAQLGDA VQLASLPPAG DILPNKTPCY ITGWGRLYTN GPLPDKLQQA RLPVVDYKHC SRWNWWGSTV KKTMVCAGGY IRSGCNGDSG GPLNCPTEDG GWQVHGVTSF VSAFGCNFIW KPTVFTRVSA FIDWIEETIA SH | |
Greater than 90% as determined by SDS-PAGE. |
10136 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |