missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Chromobox protein homolog 7 Protein
A DNA sequence encoding the Homo sapiens (Human) Chromobox protein homolog 7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5769-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_783640.1 | |
Chromobox protein homolog 7 Protein | |
Research Use Only | |
CBX7 | |
Recombinant Protein | |
E. coli | |
MELSAIGEQV FAVESIRKKR VRKGKVEYLV KWKGWPPKYS TWEPEEHILD PRLVMAYEEK EERDRASGYR KRGPKPKRLL LQRLYSMDLR SSHKAKGKEK LCFSLTCPLG SGSPEGVVKA GAPELVDKGP LVPTLPFPLR KPRKAHKYLR LSRKKFPPRG PNLESHSHRR ELFLQEPPAP DVLQAAGEWE PAAQPPEEEA DADLAEGPPP WTPALPSSEV TVTDITANSI TVTFREAQAA EGFFRDRSGK F | |
Greater than 90% as determined by SDS-PAGE. |
23492 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |