missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDKN2D / p19ink4d Protein
A DNA sequence encoding the Homo sapiens (Human) CDKN2D / p19ink4d, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5809-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001791.1 | |
CDKN2D / p19ink4d Protein | |
Research Use Only | |
CDKN2D | |
Recombinant Protein | |
E. coli | |
MLLEEVRAGD RLSGAAARGD VQEVRRLLHR ELVHPDALNR FGKTALQVMM FGSTAIALEL LKQGASPNVQ DTSGTSPVHD AARTGFLDTL KVLVEHGADV NVPDGTGALP IHLAVQEGHT AVVSFLAAES DLHRRDARGL TPLELALQRG AQDLVDILQG HMVAPL | |
Greater than 80% as determined by SDS-PAGE. |
1032 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |