missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDC26 Protein
A cDNA sequence encoding the CDC26 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11362-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CDC26 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CDC26 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 40% glycerol and 0.1M NaCl. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MLRRKPTRLE LKLDDIEEFE NIRKDLETRK KQKEDVEVVG GSDGEGAIGL SSDPKSREQM INDRIGYKPQ PKPNNRSSQF GSLEF |