missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD90 Protein
A cDNA sequence encoding the CD90 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11355-1ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD90 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
QKVTSLTACL VDQSLRLDCR HENTSSSPIQ YEFSLTRETK KHVLFGTVGV PEHTYRSRTN FTSKYNMKVL YLSAFTSKDE GTYTCALHHS GHSPPISSQN VTVLRDKLVK CHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CD90 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |