missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD35 / CR1 Protein
A DNA sequence encoding the Homo sapiens (Human) CD35 / CR1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5860-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000564.2 | |
CD35 / CR1 Protein | |
Research Use Only | |
CR1 | |
Recombinant Protein | |
E. coli | |
DPHPDRGTSF DLIGESTIRC TSDPQGNGVW SSPAPRCGIL GHCQAPDHFL FAKLKTQTNA SDFPIGTSLK YECRPEYYGR PFSITCLDNL VWSSPKDVCK RKSCKTPPDP VNGMVHVITD IQVGSRINYS CTTGHRLIGH SSAECILSGN TAHWSTKPPI CQRIPCGLPP TIANGDFIST NRENFHYGSV VTYRC | |
Greater than 90% as determined by SDS-PAGE. |
1378 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |