missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD2 / LY37 Protein
A DNA sequence encoding the Homo sapiens (Human) CD2 / LY37, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5785-ec-10ug
Informations supplémentaires : Peso : 0.01000kg
Spécification
NP_001315538.1 | |
CD2 / LY37 Protein | |
Research Use Only | |
CD2 | |
Recombinant Protein | |
E. coli | |
KEITNALETW GALGQDINLD IPSFQMSDDI DDIKWEKTSD KKKIAQFRKE KETFKEKDTY KLFKNGTLKI KHLKTDDQDI YKVSIYDTKG KNVLEKIFDL KIQERVSKPK ISWTCINTTL TCEVMNGTDP ELNLYQDGKH LKLSQRVITH KWTTSLSAKF KCTAGNKVSK ESSVEPVSCP EKGLD | |
Greater than 90% as determined by SDS-PAGE. |
914 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |