missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD160 Protein
A cDNA sequence encoding the CD160 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11302-1ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD160 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADLINITSSA SQEGTRLNLI CTVWHKKEEA EGFVVFLCKD RSGDCSPETS LKQLRLKRDP GIDGVGEISS QLMFTISQVT PLHSGTYQCC ARSQKSGIRL QGHFFSILFT ETGNYTVTGL KQRQHLEFSH NEGTLSHHHH HH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CD160 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |