missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL3 / Mip1a Protein
A DNA sequence encoding the Human CCL3 / Mip1a, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10226-ec-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002974.1 | |
CCL3 / Mip1a Protein | |
Research Use Only | |
CCL3 | |
The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 1.0-10 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
6348 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
ASLAADTPTA CCFSYTSRQI PQNFIADYFE TSSQCSKPGV IFLTKRSRQV CADPSEEWVQ KYVSDLELSA | |
Greater than 96% as determined by SDS-PAGE and HPLC. |