missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL21 / 6Ckine Protein
A DNA sequence encoding the Homo sapiens (Human) CCL21 / 6Ckine, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5773-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002980.1 | |
CCL21 / 6Ckine Protein | |
Research Use Only | |
CCL21 | |
Recombinant Protein | |
E. coli | |
SDGGAQDCCL KYSQRKIPAK VVRSYRKQEP SLGCSIPAIL FLPRKRSQAE LCADPKELWV QQLMQHLDKT PSPQKPAQGC RKDRGASKTG KKGKGSKGCK RTERSQTPKG P | |
Greater than 90% as determined by SDS-PAGE. |
6366 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |