missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL18 / PARC / MIP4 Protein
A DNA sequence encoding the Homo sapiens (Human) CCL18 / PARC / MIP4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP4989-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002979.1 | |
CCL18 / PARC / MIP4 Protein | |
Research Use Only | |
CCL18 | |
Recombinant Protein | |
Yeast | |
AQVGTNKELC CLVYTSWQIP QKFIVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA | |
Greater than 90% as determined by SDS-PAGE. |
6362 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |