missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL14 / HCC-1 / HCC-3 Protein
A DNA sequence encoding the Human CCL14 / HCC-1 / HCC-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10199-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_116738.1 | |
CCL14 / HCC-1 / HCC-3 Protein | |
Research Use Only | |
CCL14 | |
The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration of 5.0-20 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
6358 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
TESSSRGPYH PSECCFTYTT YKIPRQRIMD YYETNSQCSK PGIVFITKRG HSVCTNPSDK WVQDYIKDMK EN | |
Greater than 96% as determined by SDS-PAGE and HPLC. |