Learn More
enQuireBio™ Recombinant Human Casein kinase I isoform alpha-like Protein
A DNA sequence encoding the Homo sapiens (Human) Casein kinase I isoform alpha-like, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7904-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_660204.2 | |
Casein kinase I isoform alpha-like Protein | |
100 μg | |
Not determined. | |
Full Length of BC028723 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
122011 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
CSNK1A1L | |
Recombinant Protein | |
E. coli | |
MTNNSGSKAE LVVGGKYKLV RKIGSGSFGD VYLGITTTNG EEVAVKLESQ KVKHPQLLYE SKLYTILQGG VGIPHMHWYG QEKDNNVLVM DLLGPSLEDL FNFCSRRFTM KTVLMLADQM ISRIEYVHTK NFLHRDIKPD NFLMGTGRHC NKLFLIDFGL AKKYRDNRTR QHIPYREDKH LIGTVRYASI NAHLGIEQSR RDDMESLGYV FMYFNRTSLP WQGLKAMTKK QKYEKISEKK MSTPVEVLCK GFPAEFAMYL NYCRGLRFEE VPDYMYLRQL FRILFRTLNH QYDYTFDWTM LKQKAAQQAA SSSGQGQQAQ TQTGKQTEKN KNNVKDN |