missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Carboxypeptidase A6 Protein
A DNA sequence encoding the Homo sapiens (Human) Carboxypeptidase A6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5857-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_065094.3 | |
Carboxypeptidase A6 Protein | |
Research Use Only | |
CPA6 | |
Recombinant Protein | |
E. coli | |
SLSGYNYEVY HSLEEIQNWM HHLNKTHSGL IHMFSIGRSY EGRSLFILKL GRRSRLKRAV WIDCGIHARE WIGPAFCQWF VKEALLTYKS DPAMRKMLNH LYFYIMPVFN VDGYHFSWTN DRFWRKTRSR NSRFRCRGVD ANRNWKVKWC DEGASMHPCD DTYCGPFPES EPEVKAVANF LRKHRKHIRA YLSFHAYAQM LLYPYSYKYA TIPNFRCVES AAYKAVNALQ SVYGVRYRYG PASTTLYVSS GSSMDWAYKN GIPYAFAFEL RDTGYFGFLL PEMLIKPTCT ETMLAVKNIT MHLLKKCP | |
Greater than 90% as determined by SDS-PAGE. |
57094 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |