missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Carbonic Anhydrase XII / CA12 Protein
A DNA sequence encoding the Homo sapiens (Human) Carbonic Anhydrase XII / CA12, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5742-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001209.1 | |
Carbonic Anhydrase XII / CA12 Protein | |
Research Use Only | |
CA12 | |
Recombinant Protein | |
E. coli | |
APVNGSKWTY FGPDGENSWS KKYPSCGGLL QSPIDLHSDI LQYDASLTPL EFQGYNLSAN KQFLLTNNGH SVKLNLPSDM HIQGLQSRYS ATQLHLHWGN PNDPHGSEHT VSGQHFAAEL HIVHYNSDLY PDASTASNKS EGLAVLAVLI EMGSFNPSYD KIFSHLQHVK YKGQEAFVPG FNIEELLPER TAEYYRYRGS LTTPPCNPTV LWTVFRNPVQ ISQEQLLALE TALYCTHMDD PSPREMINNF RQVQKFDERL VYTSFSQVQV CTAAGLS | |
Greater than 90% as determined by SDS-PAGE. |
771 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |