missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cancer / testis antigen 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Cancer/testis antigen 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5883-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_066274.2 | |
Cancer / testis antigen 2 Protein | |
Research Use Only | |
CTAG2 | |
Recombinant Protein | |
E. coli | |
MQAEGQGTGG STGDADGPGG PGIPDGPGGN AGGPGEAGAT GGRGPRGAGA ARASGPRGGA PRGPHGGAAS AQDGRCPCGA RRPDSRLLQL HITMPFSSPM EAELVRRILS RDAAPLPRPG AVLKDFTVSG NLLFMSVRDQ DREGAGRMRV VGWGLGSASP EGQKARDLRT PKHKVSEQRP GTPGPPPPEG AQGDGCRGVA FNVMFSAPHI | |
Greater than 90% as determined by SDS-PAGE. |
30848 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |