missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CALR3 Protein

A cDNA sequence encoding the CALR3 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11264-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15936658

  • 161.00€ / 20 microgramos


Fecha estimada de envío
20-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

CALR3 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
CALR3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea, and 10% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MTVYFQEEFL DGEHWRNRWL QSTNDSRFGH FRLSSGKFYG HKEKDKGLQT TQNGRFYAIS ARFKPFSNKG KTLVIQYTVK HEQKMDCGGG YIKVFPADID QKNLNGKSQY YIMFGPDICG FDIKKVHVIL HFKNKYHENK KLIRCKVDGF THLYTLILRP DLSYDVKIDG QSIESGSIEY DWNLTSLKKE TSPAESKDWE QTKDNKAQDW EKHFLDASTS KQSDWNGDLD GDWPAPMLQK PPYQDGLKPE GIHKDVWLHR KMKNTDYLTQ YDLSEFENIG AIGLELWQVR SGTIFDNFLI TDDEEYADNF GKATWGETKG PEREMDAIQA KEEMKKAREE EEEELLSGKI NRHEHYFNQF HRRNEL
Videos
Documentos