missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calponin-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Calponin-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5849-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001272984.1 | |
Calponin-3 Protein | |
Research Use Only | |
CNN3 | |
Recombinant Protein | |
E. coli | |
THFNKGPSYG LSAEVKNKIA SKYDHQAEED LRNWIEEVTG MSIGPNFQLG LKDGIILCEL INKLQPGSVK KVNESSLNWP QLENIGNFIK AIQAYGMKPH DIFEANDLFE NGNMTQVQTT LVALAGLAKT KGFHTTIDIG VKYAEKQTRR FDEGKLKAGQ SVIGLQMGTN KCASQAGMTA YGTRRHLYDP KMQTDKPFDQ TTISLQMGTN KGASQAGMLA PGTRRDIYDQ KLTLQPVDNS TISLQMGTNK VASQKGMSVY GLGRQVYDPK YCAAPTEPVI HNGSQGTGTN GSEISDSDYQ AEYPDEYHGE YQDDYPRDYQ YSDQGIDY | |
Greater than 90% as determined by SDS-PAGE. |
1266 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |