missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calpain small subunit 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Calpain small subunit 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5758-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001003962.1 | |
Calpain small subunit 1 Protein | |
Research Use Only | |
CAPNS1 | |
Recombinant Protein | |
E. coli | |
MFLVNSFLKG GGGGGGGGGG LGGGLGNVLG GLISGAGGGG GGGGGGGGGG GGGGGGTAMR ILGGVISAIS EAAAQYNPEP PPPRTHYSNI EANESEEVRQ FRRLFAQLAG DDMEVSATEL MNILNKVVTR HPDLKTDGFG IDTCRSMVAV MDSDTTGKLG FEEFKYLWNN IKRWQAIYKQ FDTDRSGTIC SSELPGAFEA AGFHLNEHLY NMIIRRYSDE SGNMDFDNFI SCLVRLDAMF RAFKSLDKDG TGQIQVNIQE WLQLTMYS | |
Greater than 90% as determined by SDS-PAGE. |
826 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |