missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CA13 Protein
A cDNA sequence encoding the CA13 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10542-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
377677 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 2,500 pmol/min/ug, and is defined as the amount of enzyme that hydrolyze 1.0 pmole of 4-nitrophenyl acetate to 4-nitrophenol per minute at pH 7.5 at 37C. | |
Human | |
His | |
CA13 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4),10% glycerol and 1mM DTT. |
CA13 Protein | |
Research Use Only | |
CA13 | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSP MGSMSRLSWG YREHNGPIHW KEFFPIADGD QQSPIEIKTK EVKYDSSLRP LSIKYDPSSA KIISNSGHSF NVDFDDTENK SVLRGGPLTG SYRLRQVHLH WGSADDHGSE HIVDGVSYAA ELHVVHWNSD KYPSFVEAAH EPDGLAVLGV FLQIGEPNSQ LQKITDTLDS IKEKGKQTRF TNFDLLSLLP PSWDYWTYPG SLTVPPLLES VTWIVLKQPI NISSQQLAKF RSLLCTAEGE AAAFLVSNHR PPQPLKGRKV RASFH | |
Greater than 90.0% as determined by SDS-PAGE. |