missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C5a / Complement 5a Protein
A DNA sequence encoding the Homo sapiens (Human) C5a / Complement 5a, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP5737-ma-20ug
Weitere Details : Peso : 0.01000kg
Spezifikation
NP_001304092.1 | |
C5a / Complement 5a Protein | |
Research Use Only | |
C5 | |
Recombinant Protein | |
Mammal | |
TLQKKIEEIA AKYKHSVVKK CCYDGACVNN DETCEQRAAR ISLGPRCIKA FTECCVVASQ LRANISHKDM QLGR | |
Greater than 90% as determined by SDS-PAGE. |
727 | |
20 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |