missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C1QTNF1 Protein
A cDNA sequence encoding the C1QTNF1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11230-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
C1QTNF1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS RVPHVQGEQQ EWEGTEELPS PPDHAERAEE QHEKYRPSQD QGLPASRCLR CCDPGTSMYP ATAVPQINIT ILKGEKGDRG DRGLQGKYGK TGSAGARGHT GPKGQKGSMG APGERCKSHY AAFSVGRKKP MHSNHYYQTV IFDTEFVNLY DHFNMFTGKF YCYVPGLYFF SLNVHTWNQK ETYLHIMKNE EEVVILFAQV GDRSIMQSQS LMLELREQDQ VWVRLYKGER ENAIFSEELD TYITFSGYLVKHATEP | |
Greater than 95% as determined by SDS PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Human C1QTNF1 was lyophilized from 50mM Acetate Buffer pH 4. |